missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3',4',5'-Trimethoxyacetophenone, 99%

Manufacturer:  Alfa Aesar™ L0643906

 View more versions of this product

Catalog No. AAL0643906



Description

Description

Applications
3′,4′,5′-Trimethoxyacetophenone is used in agrochemical, pharmaceutical and dyestuff field .

Solubility
Slightly soluble in water.

Notes
Incompatible materials are Strong oxidizing agents, Strong bases. Store away from strong oxidizing agents and stong bases. Avoid excessive heat and light exposure.
Specifications

Specifications

3',4',5'-Trimethoxyacetophenone
79°C to 83°C
1136-86-3
MFCD00008738
1-3,4,5-trimethoxyphenyl ethan-1-one
CC(=O)C1=CC(=C(C(=C1)OC)OC)OC
210.229
CHEBI:86547
99%
173°C to 174°C (10mmHg)
5g
C11H14O4
1463133
VUGQIIQFXCXZJU-UHFFFAOYSA-N
1-(3,4,5-trimethoxyphenyl)ethanone
14345
210.23

Chemical Identifiers

1136-86-3
210.229
VUGQIIQFXCXZJU-UHFFFAOYSA-N
14345
1-(3,4,5-trimethoxyphenyl)ethanone
C11H14O4
MFCD00008738
1-3,4,5-trimethoxyphenyl ethan-1-one
CHEBI:86547
CC(=O)C1=CC(=C(C(=C1)OC)OC)OC
Documents