Please login to your online account to display your discounted pricing

3,4,5-Trifluorophenylboronic acid, 95+%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  368190050

 View more versions of this product

Catalog No. AC368190050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Infrared Spectrum Authentic
Packaging Glass bottle
Color Tan
Quantity 5g
Chemical Name or Material 3, 4, 5-Trifluorophenylboronic acid
CAS 143418-49-9
CAS Min % 95.0
CAS Max % 100.0
Physical Form Tan powder
Assay 95.0% min.
Assay Percent Range 95+%
Molecular Formula C6H4BF3O2
Formula Weight 175.9g/mol
MDL Number MFCD02093069
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Toxic to aquatic life with long lasting effects.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 3, 4, 5-Trifluorophenylboronic acid
CAS 143418-49-9
CAS Min % 95.0
CAS Max % 100.0
Assay 95.0% min.
Assay Percent Range 95+%
Molecular Formula C6H4BF3O2
Formula Weight 175.9g/mol
MDL Number MFCD02093069
Accessories

GHS Signal Word: Warning

GHS P Statement:
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Toxic to aquatic life with long lasting effects.
Causes serious eye irritation.

SDS & Safety Documents