Please login to your online account to display your discounted pricing

3,4,5-Trifluorophenylboronic acid, 95+%, ACROS Organics™


Quantity 250mg
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC368192500

 View more versions of this product

Catalog No. AC368192500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥95.0%
Physical Form Powder
Formula Weight 175.9
Quantity 250mg
Color Tan
CAS Min % 95.0
CAS Max % 100.0
Packaging Glass bottle
Assay Percent Range 95+%
MDL Number MFCD02093069
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Toxic to aquatic life with long lasting effects.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 3, 4, 5-Trifluorophenylboronic acid
Molecular Formula C6H4BF3O2
CAS 143418-49-9
Health Hazard 1 GHS Signal Word: Warning

Specifications

Chemical Name or Material 3, 4, 5-Trifluorophenylboronic acid
CAS 143418-49-9
CAS Min % 95.0
CAS Max % 100.0
Assay ≥95.0%
Assay Percent Range 95+%
Molecular Formula C6H4BF3O2
Formula Weight 175.9
MDL Number MFCD02093069
Accessories

GHS Signal Word: Warning

GHS P Statement:
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Toxic to aquatic life with long lasting effects.
Causes serious eye irritation.

SDS & Safety Documents