missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,4,5-Trifluorophenol, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC442580010

 View more versions of this product

Catalog No. AC442580010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

3, 4, 5-Trifluorophenol
Glass Bottle
50.0°C to 51.0°C
99627-05-1
100.0
C6H3F3O
ZRTWIJKGTUGZJY-UHFFFAOYSA-N
3,4,5-trifluorophenol
2777943
Crystals or Low Melting Solid
Authentic
Colorless to Yellow
1g
97.5
97.5% min. (GC)
3 , 4 , 5-trifluorophenol, 3,4,5-trifluoro-phenol, 3,4,5-trifluoropheno, 3,4,5-trifluorophenol, acmc-209sdi, ksc486m6r, phenol, 3,4,5-trifluoro, phenol, 3,4,5-trifluoro-9ci, pubchem2316
C1=C(C=C(C(=C1F)F)F)O
148.09
148.09
98%

Chemical Identifiers

99627-05-1
148.09
3 , 4 , 5-trifluorophenol, 3,4,5-trifluoro-phenol, 3,4,5-trifluoropheno, 3,4,5-trifluorophenol, acmc-209sdi, ksc486m6r, phenol, 3,4,5-trifluoro, phenol, 3,4,5-trifluoro-9ci, pubchem2316
3,4,5-trifluorophenol
C6H3F3O
ZRTWIJKGTUGZJY-UHFFFAOYSA-N
2777943
C1=C(C=C(C(=C1F)F)F)O
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with

GHS Signal Word: Danger

SDS Documents