Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,4,5-Trifluorobenzyl alcohol, 97%

$101.45 - $410.99

Chemical Identifiers

Chemical Name or Material 3,4,5-Trifluorobenzyl alcohol
CAS 220227-37-2
Molecular Formula C7H5F3O
Formula Weight 162.11
MDL Number MFCD00083527
Beilstein 8681069
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2461703

 
alfa aesar™
B2461703
1g Each for $101.45

AAB2461706

 
alfa aesar™
B2461706
5g Each for $410.99
Description & Specifications

Specifications

Formula Weight 162.11
Chemical Name or Material 3,4,5-Trifluorobenzyl alcohol
Assay 97%
Molecular Formula C7H5F3O
MDL Number MFCD00083527
CAS 220227-37-2
Beilstein 8681069
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Flash Point 98°C (208°F)
Boiling Point 203° to 204°C
Quantity 1g

Applications
3,4,5-Trifluorobenzyl alcohol is used as chemical reagent.

Solubility
Soluble in water 100 g/L.

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.