Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,4,5-Trifluorobenzophenone, 97%

$45.51 - $135.30

Chemical Identifiers

Chemical Name or Material 3,4,5-Trifluorobenzophenone
CAS 70028-88-5
Molecular Formula C13H7F3O
Formula Weight 236.19
MDL Number MFCD00083525
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2672703

  SDS
alfa aesar™
H2672703
1g Each for $45.51

AAH2672706

  SDS
alfa aesar™
H2672706
5g Each for $135.30
Description & Specifications

Specifications

Formula Weight 236.19
Chemical Name or Material 3,4,5-Trifluorobenzophenone
Assay 97%
Molecular Formula C13H7F3O
MDL Number MFCD00083525
CAS 70028-88-5
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.