missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,4,5-Trifluorobenzoic acid, 97%

$89.45 - $326.93

Chemical Identifiers

CAS 121602-93-5
Molecular Formula C7H3F3O2
Molecular Weight (g/mol) 176.094
MDL Number MFCD00074962
InChI Key VJMYKESYFHYUEQ-UHFFFAOYSA-N
Synonym 3,4,5-trifluorobenzoicacid, benzoic acid, 3,4,5-trifluoro, 3,4,5-trifluoro benzoic acid, 3,4,5-trifluoro-benzoic acid, acmc-209aen, ksc177i9t, rarechem al bo 0682, 3,4,5-trifluorobenzoyl alcohol, buttpark 30\01-46
PubChem CID 853154
IUPAC Name 3,4,5-trifluorobenzoic acid
SMILES C1=C(C=C(C(=C1F)F)F)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1893903 View Documents Alfa Aesar™
L1893903
1g Each for $89.45
 
AAL1893906 View Documents Alfa Aesar™
L1893906
5g Each for $326.93
 
Specifications

Chemical Identifiers

121602-93-5
176.094
VJMYKESYFHYUEQ-UHFFFAOYSA-N
853154
C1=C(C=C(C(=C1F)F)F)C(=O)O
C7H3F3O2
MFCD00074962
3,4,5-trifluorobenzoicacid, benzoic acid, 3,4,5-trifluoro, 3,4,5-trifluoro benzoic acid, 3,4,5-trifluoro-benzoic acid, acmc-209aen, ksc177i9t, rarechem al bo 0682, 3,4,5-trifluorobenzoyl alcohol, buttpark 30\01-46
3,4,5-trifluorobenzoic acid

Specifications

97°C to 99°C
121602-93-5
MFCD00074962
3,4,5-trifluorobenzoicacid, benzoic acid, 3,4,5-trifluoro, 3,4,5-trifluoro benzoic acid, 3,4,5-trifluoro-benzoic acid, acmc-209aen, ksc177i9t, rarechem al bo 0682, 3,4,5-trifluorobenzoyl alcohol, buttpark 30\01-46
C1=C(C=C(C(=C1F)F)F)C(=O)O
176.094
176.1
3,4,5-Trifluorobenzoic acid
1g
C7H3F3O2
7476737
VJMYKESYFHYUEQ-UHFFFAOYSA-N
3,4,5-trifluorobenzoic acid
853154
97%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning