Please login to your online account to display your discounted pricing

3,4,5-trifluorobenzenesulfonyl Chloride 98%, ACROS Organics™

$627.16 - $627.16

Chemical Identifiers

Chemical Name or Material 3, 4, 5-Trifluorobenzenesulfonyl chloride
CAS 351003-43-5
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C6 H2 Cl F3 O2 S
Linear Formula F3C6H2SO2Cl
Formula Weight 230.59
MDL Number MFCD03094559
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC383240050

  SDS
thermo fisher scientific
AC383240050
Glass bottle 5g Each for $627.16
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Liquid
Formula Weight 230.59
Quantity 5g
Color Clear colorless
Density 1.6430g/mL
CAS Max % 100.0
Linear Formula F3C6H2SO2Cl
Infrared Spectrum Authentic
Refractive Index 1.4960 to 1.4980 (20°C, 589nm)
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD03094559
Packaging Glass bottle
Chemical Name or Material 3, 4, 5-Trifluorobenzenesulfonyl chloride
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
Contact with water liberates toxic gas.
CAS Min % 97.5
Molecular Formula C6 H2 Cl F3 O2 S
CAS 351003-43-5
Health Hazard 1 GHS Signal Word: Danger
Assay Percent Range 98%
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
Contact with water liberates toxic gas.