Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,4,5-Trifluorobenzenesulfonyl chloride, 98%

Chemical Identifiers

Chemical Name or Material 3,4,5-Trifluorobenzenesulfonyl chloride
CAS 351003-43-5
Molecular Formula C6H2ClF3O2S
Formula Weight 230.59
MDL Number MFCD03094559
UN Number UN3265
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1990506

 
alfa aesar™
L1990506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 230.59
Assay 98%
MDL Number MFCD03094559
UN Number UN3265
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
Flash Point >100°C (212°F)
Chemical Name or Material 3,4,5-Trifluorobenzenesulfonyl chloride
CAS 351003-43-5
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Molecular Formula C6H2ClF3O2S
Boiling Point ≈230°C
Quantity 5g
Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.