missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,4,5-Trifluorobenzenesulfonamide, 97%

$121.51 - $402.50

Chemical Identifiers

CAS 913472-55-6
Molecular Formula C6H4F3NO2S
Molecular Weight (g/mol) 211.158
MDL Number MFCD09800668
InChI Key BIAGBHUIRSMWRI-UHFFFAOYSA-N
Synonym 3,4,5-trifluorobenzene-1-sulfonamide
PubChem CID 28306934
IUPAC Name 3,4,5-trifluorobenzenesulfonamide
SMILES C1=C(C=C(C(=C1F)F)F)S(=O)(=O)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH3196403 View Documents Alfa Aesar™
H3196403
1g Each for $121.51
 
AAH3196406 View Documents Alfa Aesar™
H3196406
5g Each for $402.50
 
Specifications

Chemical Identifiers

913472-55-6
211.158
BIAGBHUIRSMWRI-UHFFFAOYSA-N
28306934
C1=C(C=C(C(=C1F)F)F)S(=O)(=O)N
C6H4F3NO2S
MFCD09800668
3,4,5-trifluorobenzene-1-sulfonamide
3,4,5-trifluorobenzenesulfonamide

Specifications

124°C to 126°C
913472-55-6
MFCD09800668
BIAGBHUIRSMWRI-UHFFFAOYSA-N
3,4,5-trifluorobenzenesulfonamide
28306934
97%
1g
C6H4F3NO2S
3,4,5-trifluorobenzene-1-sulfonamide
C1=C(C=C(C(=C1F)F)F)S(=O)(=O)N
211.158
211.16
3,4,5-Trifluorobenzenesulfonamide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning