Please login to your online account to display your discounted pricing

3,4,5-trifluorobenzaldehyde 98%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC384210050

 View more versions of this product

Catalog No. AC384210050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Liquid After Melting
Formula Weight 160.09
Quantity 5g
Color Clear colorless to slightly yellow
Density 1.4200g/mL
CAS Max % 100.0
Packaging Glass bottle
CAS 132123-54-7
Health Hazard 1 GHS Signal Word: Warning
Chemical Name or Material 3, 4, 5-Trifluorobenzaldehyde
Infrared Spectrum Authentic
Assay Percent Range 98%
Linear Formula F3C6H2CHO
MDL Number MFCD00083523
CAS Min % 97.5
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Harmful if swallowed.
Toxic to aquatic life with long lasting effects.
Health Hazard 3 GHS P Statement:
Wear eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Molecular Formula C7H3F3O

Specifications

Chemical Name or Material 3, 4, 5-Trifluorobenzaldehyde
CAS 132123-54-7
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C7H3F3O
Linear Formula F3C6H2CHO
Formula Weight 160.09
MDL Number MFCD00083523
Accessories

GHS Signal Word: Warning

GHS P Statement:
Wear eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Harmful if swallowed.
Toxic to aquatic life with long lasting effects.

SDS & Safety Documents