Please login to your online account to display your discounted pricing

3,4,5-trifluorobenzaldehyde 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3, 4, 5-Trifluorobenzaldehyde
CAS 132123-54-7
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C7H3F3O
Linear Formula F3C6H2CHO
Formula Weight 160.09
MDL Number MFCD00083523
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC384210010

  SDS
thermo fisher scientific
AC384210010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC384210050

  SDS
thermo fisher scientific
AC384210050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Formula Weight 160.09
Physical Form Liquid After Melting
Quantity 1g
Color Clear colorless to slightly yellow
Density 1.4200g/mL
CAS Max % 100.0
Health Hazard 1 GHS Signal Word: Warning
MDL Number MFCD00083523
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Harmful if swallowed.
Toxic to aquatic life with long lasting effects.
CAS Min % 97.5
Infrared Spectrum Authentic
Packaging Glass bottle
Linear Formula F3C6H2CHO
Assay Percent Range 98%
CAS 132123-54-7
Chemical Name or Material 3, 4, 5-Trifluorobenzaldehyde
Molecular Formula C7H3F3O
Health Hazard 3 GHS P Statement:
Wear eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Wear eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Harmful if swallowed.
Toxic to aquatic life with long lasting effects.