Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,4,5-Trifluorobenzaldehyde, 98%

$71.12 - $215.47

Chemical Identifiers

Chemical Name or Material 3,4,5-Trifluorobenzaldehyde
CAS 132123-54-7
Molecular Formula C7H3F3O
Formula Weight 160.1
MDL Number MFCD00083523
Beilstein 7805841
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2094303

  SDS
alfa aesar™
B2094303
1g Each for $71.12

AAB2094306

  SDS
alfa aesar™
B2094306
5g Each for $215.47
Description & Specifications

Specifications

Formula Weight 160.1
Boiling Point 174°C
Beilstein 7805841
MDL Number MFCD00083523
Health Hazard 1 Warning
Quantity 1g
Assay 98%
Molecular Formula C7H3F3O
Melting Point 24° to 26°C
Chemical Name or Material 3,4,5-Trifluorobenzaldehyde
CAS 132123-54-7
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Flash Point 62°C (144°F)
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.