Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,4,5-Trifluoroanisole, 97%

Chemical Identifiers

Chemical Name or Material 3,4,5-Trifluoroanisole
CAS 203245-17-4
Molecular Formula C7H5F3O
Formula Weight 162.11
MDL Number MFCD00042397
UN Number UN3271
Synonym 5-Methoxy-1,2,3-trifluorobenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2642003

  SDS
alfa aesar™
H2642003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH2642006

  SDS
alfa aesar™
H2642006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 162.11
Chemical Name or Material 3,4,5-Trifluoroanisole
Assay 97%
Molecular Formula C7H5F3O
MDL Number MFCD00042397
CAS 203245-17-4
UN Number UN3271
Synonym 5-Methoxy-1,2,3-trifluorobenzene
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H226
Flammable liquid and vapour.
Health Hazard 3 GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ETHERS, N.O.S.
Flash Point 52°C (126°F)
Boiling Point 39°C (1mmHg)
Melting Point ≈20°C
Quantity 1g

Applications
3,4,5-Trifluoroanisole is used as a pharmaceutical intermediate.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226
Flammable liquid and vapour.