missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3',4',5'-Trifluoroacetophenone, 97%

Manufacturer:  Alfa Aesar™ L1918706

 View more versions of this product

Catalog No. AAL1918706



Specifications

Specifications

3',4',5'-Trifluoroacetophenone
5g
C8H5F3O
1-3,4,5-trifluoro-phenyl-ethanone
CC(=O)C1=CC(=C(C(=C1)F)F)F
174.122
174.12
>100°C (212°F)
220141-73-1
MFCD00083533
KXVIYAHOUAMVJX-UHFFFAOYSA-N
1-(3,4,5-trifluorophenyl)ethanone
2776907
97%

Chemical Identifiers

220141-73-1
174.122
KXVIYAHOUAMVJX-UHFFFAOYSA-N
2776907
CC(=O)C1=CC(=C(C(=C1)F)F)F
C8H5F3O
MFCD00083533
1-3,4,5-trifluoro-phenyl-ethanone
1-(3,4,5-trifluorophenyl)ethanone
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents