Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,4,5-Triethoxybenzoic acid, 98+%

$89.80 - $399.30

Chemical Identifiers

Chemical Name or Material 3,4,5-Triethoxybenzoic acid
CAS 6970-19-0
Molecular Formula C13H18O5
Formula Weight 254.29
MDL Number MFCD00002505
Beilstein 2659063
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1415722

 
alfa aesar™
A1415722
100g Each for $89.80

AAA1415736

 
alfa aesar™
A1415736
500g Each for $399.30

AAA1415714

 
alfa aesar™
A1415714
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material 3,4,5-Triethoxybenzoic acid
Quantity 100g
CAS 6970-19-0
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay ≥98%
Molecular Formula C13H18O5
Formula Weight 254.29
MDL Number MFCD00002505
Beilstein 2659063
Melting Point 110° to 113°C
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.