missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,4,5-Trichlorothiophene-2-carboxylic acid, 98+%

$52.58 - $168.09

Chemical Identifiers

CAS 26020-48-4
Molecular Formula C5HCl3O2S
Molecular Weight (g/mol) 231.471
MDL Number MFCD00092429
InChI Key FVFYTPFJDMYLJS-UHFFFAOYSA-N
Synonym 3,4,5-trichloro-2-thiophenecarboxylic acid, trichlorothiophene-2-carboxylic acid, acmc-1cbhi, 3,4,5-trichlorothio-phene-2-carboxylic acid, 3,4,5-tris chloranyl thiophene-2-carboxylic acid
PubChem CID 4168584
IUPAC Name 3,4,5-trichlorothiophene-2-carboxylic acid
SMILES C1(=C(SC(=C1Cl)Cl)C(=O)O)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1775003 View Documents Alfa Aesar™
L1775003
1g Each for $52.58
 
AAL1775006 View Documents Alfa Aesar™
L1775006
5g Each for $168.09
 
Description

Description

Applications
It is a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store at room temperature. Keep away from strong oxidizing agents. Ensure proper ventilation.
Specifications

Chemical Identifiers

26020-48-4
231.471
FVFYTPFJDMYLJS-UHFFFAOYSA-N
4168584
C1(=C(SC(=C1Cl)Cl)C(=O)O)Cl
C5HCl3O2S
MFCD00092429
3,4,5-trichloro-2-thiophenecarboxylic acid, trichlorothiophene-2-carboxylic acid, acmc-1cbhi, 3,4,5-trichlorothio-phene-2-carboxylic acid, 3,4,5-tris chloranyl thiophene-2-carboxylic acid
3,4,5-trichlorothiophene-2-carboxylic acid

Specifications

226°C to 228°C
26020-48-4
MFCD00092429
3,4,5-trichloro-2-thiophenecarboxylic acid, trichlorothiophene-2-carboxylic acid, acmc-1cbhi, 3,4,5-trichlorothio-phene-2-carboxylic acid, 3,4,5-tris chloranyl thiophene-2-carboxylic acid
C1(=C(SC(=C1Cl)Cl)C(=O)O)Cl
231.471
231.49
3,4,5-Trichlorothiophene-2-carboxylic acid
1g
C5HCl3O2S
155175
FVFYTPFJDMYLJS-UHFFFAOYSA-N
3,4,5-trichlorothiophene-2-carboxylic acid
4168584
≥98%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning