missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,4,5-Trichlorothiophene-2-carbonyl chloride, 97%

$61.82 - $189.68

Chemical Identifiers

CAS 24422-15-9
Molecular Formula C5Cl4OS
Molecular Weight (g/mol) 249.914
MDL Number MFCD02093952
InChI Key ASESVIHJSOBINF-UHFFFAOYSA-N
Synonym 3,4,5-trichloro-2-thiophenecarbonyl chloride, trichlorothiophene-2-carbonyl chloride, acmc-20angk, 3,4,5-trichlorothiophene-2-carbonylchloride, 2-thiophenecarbonylchloride, 3,4,5-trichloro, 3,4,5-tris chloranyl thiophene-2-carbonyl chloride
PubChem CID 7010308
IUPAC Name 3,4,5-trichlorothiophene-2-carbonyl chloride
SMILES C1(=C(SC(=C1Cl)Cl)C(=O)Cl)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1770403 View Documents Alfa Aesar™
L1770403
1g Each for $61.82
 
AAL1770406 View Documents Alfa Aesar™
L1770406
5g Each for $189.68
 
Specifications

Chemical Identifiers

24422-15-9
249.914
ASESVIHJSOBINF-UHFFFAOYSA-N
7010308
C1(=C(SC(=C1Cl)Cl)C(=O)Cl)Cl
C5Cl4OS
MFCD02093952
3,4,5-trichloro-2-thiophenecarbonyl chloride, trichlorothiophene-2-carbonyl chloride, acmc-20angk, 3,4,5-trichlorothiophene-2-carbonylchloride, 2-thiophenecarbonylchloride, 3,4,5-trichloro, 3,4,5-tris chloranyl thiophene-2-carbonyl chloride
3,4,5-trichlorothiophene-2-carbonyl chloride

Specifications

124°C to 125°C (10mmHg)
1g
C5Cl4OS
UN3261
3,4,5-trichloro-2-thiophenecarbonyl chloride, trichlorothiophene-2-carbonyl chloride, acmc-20angk, 3,4,5-trichlorothiophene-2-carbonylchloride, 2-thiophenecarbonylchloride, 3,4,5-trichloro, 3,4,5-tris chloranyl thiophene-2-carbonyl chloride
C1(=C(SC(=C1Cl)Cl)C(=O)Cl)Cl
249.914
249.93
3,4,5-Trichlorothiophene-2-carbonyl chloride
38°C to 40°C
24422-15-9
MFCD02093952
1427060
ASESVIHJSOBINF-UHFFFAOYSA-N
3,4,5-trichlorothiophene-2-carbonyl chloride
7010308
97%
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.