Learn More
3,4,5-Trichloropyridazine, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 160590050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
3,4,5-Trichloropyridazine | |
117.0°C to 118.0°C (14.0 mmHg) | |
97.5% min. (HPLC) | |
5g | |
GBAOOJAWDCNOGO-UHFFFAOYSA-N | |
3,4,5-trichloropyridazine | |
70111 | |
98% |
14161-11-6 | |
Authentic | |
C4HCl3N2 | |
pyridazine, 3,4,5-trichloro, 3,4,5-trichloro-pyridazine, trichloropyridazine, ksc493s5t, gbaoojawdcnogo-uhfffaoysa, pyridazine,3,4,5-trichloro, 3,4,5-trichloro-1,2-diazine | |
C1=C(C(=C(N=N1)Cl)Cl)Cl | |
183.42 | |
183.42 |
Chemical Identifiers
14161-11-6 | |
183.42 | |
pyridazine, 3,4,5-trichloro, 3,4,5-trichloro-pyridazine, trichloropyridazine, ksc493s5t, gbaoojawdcnogo-uhfffaoysa, pyridazine,3,4,5-trichloro, 3,4,5-trichloro-1,2-diazine | |
3,4,5-trichloropyridazine |
C4HCl3N2 | |
GBAOOJAWDCNOGO-UHFFFAOYSA-N | |
70111 | |
C1=C(C(=C(N=N1)Cl)Cl)Cl |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
GHS Signal Word: Danger
EINECSNumber : 238-006-2
RUO â Research Use Only