missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,4,5-Trichloropyridazine, 98%, Acros Organics™

Manufacturer:  Thermo Fisher Scientific AC160590010

 View more versions of this product

Catalog No. AC160590010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

3,4,5-Trichloropyridazine
Glass Bottle
54.0°C to 61.0°C
14161-11-6
C4HCl3N2
GBAOOJAWDCNOGO-UHFFFAOYSA-N
3,4,5-trichloropyridazine
70111
98%
Authentic
117.0°C to 118.0°C (14.0 mmHg)
1g
97.5% min. (HPLC)
3,4,5-trichloro-1,2-diazine
C1=C(C(=C(N=N1)Cl)Cl)Cl
183.42
183.42

Chemical Identifiers

14161-11-6
183.42
3,4,5-trichloro-1,2-diazine
3,4,5-trichloropyridazine
C4HCl3N2
GBAOOJAWDCNOGO-UHFFFAOYSA-N
70111
C1=C(C(=C(N=N1)Cl)Cl)Cl
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS Signal Word: Danger

EINECSNumber : 238-006-2

Documents