missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,4,5-Trichlorobenzotrifluoride, TCI America™

Manufacturer:  TCI America T228125G

 View more versions of this product

Catalog No. T228125G



Specifications

Specifications

3,4,5-Trichlorobenzotrifluoride
Colorless
25g
C7H2Cl3F3
FBKFIAIRSQOXJR-UHFFFAOYSA-N
1,2,3-trichloro-5-(trifluoromethyl)benzene
2733399
Clear Liquid at 20°C
113°C
-6°C
50594-82-6
1,2,3-trichloro-5-trifluoromethyl benzene, 3,4,5-trichlorobenzotrifluoride, 3,4,5-trichlorotrifluoromethylbenzene, 3,4,5-trichloro-alpha,alpha,alpha-trifluorotoluene, 3,4,5-trichloro-a,a,a-trifluorotoluene, benzene, 1,2,3-trichloro-5-trifluoromethyl, 3,4,5-trichlorobenzotrifluoride, pubchem19737, ksc914i9j, 3,4,5-trichloro benzotrifluoride
C1=C(C=C(C(=C1Cl)Cl)Cl)C(F)(F)F
249.438
249.44
≥98.0% (GC)

Chemical Identifiers

50594-82-6
249.438
1,2,3-trichloro-5-trifluoromethyl benzene, 3,4,5-trichlorobenzotrifluoride, 3,4,5-trichlorotrifluoromethylbenzene, 3,4,5-trichloro-alpha,alpha,alpha-trifluorotoluene, 3,4,5-trichloro-a,a,a-trifluorotoluene, benzene, 1,2,3-trichloro-5-trifluoromethyl, 3,4,5-trichlorobenzotrifluoride, pubchem19737, ksc914i9j, 3,4,5-trichloro benzotrifluoride
1,2,3-trichloro-5-(trifluoromethyl)benzene
C7H2Cl3F3
FBKFIAIRSQOXJR-UHFFFAOYSA-N
2733399
C1=C(C=C(C(=C1Cl)Cl)Cl)C(F)(F)F
Safety and Handling

Safety and Handling

TSCA : No

Documents