Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,4,5-Trichlorobenzotrifluoride, 97%

$30.45 - $30.45

Chemical Identifiers

Chemical Name or Material 3,4,5-Trichlorobenzotrifluoride
CAS 50594-82-6
Molecular Formula C7H2Cl3F3
Formula Weight 249.46
MDL Number MFCD00035985
Beilstein 2212413
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1419503

  SDS
alfa aesar™
L1419503
1g Each for $30.45
Description & Specifications

Specifications

Formula Weight 249.46
MDL Number MFCD00035985
CAS 50594-82-6
Chemical Name or Material 3,4,5-Trichlorobenzotrifluoride
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Assay 97%
Molecular Formula C7H2Cl3F3
Quantity 1g
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Health Hazard 1 Warning
Melting Point −10°C
Beilstein 2212413
Boiling Point 200° to 202°C
Flash Point 98°C (208°F)
Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.