Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,4,5-Tribromopyridine, 97%

$105.76 - $294.00

Chemical Identifiers

Chemical Name or Material 3,4,5-Tribromopyridine
CAS 2457-48-9
Molecular Formula C5H2Br3N
Formula Weight 315.79
MDL Number MFCD01646053
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH31426MD

 
alfa aesar™
H31426MD
250mg Each for $105.76

AAH3142603

 
alfa aesar™
H3142603
1g Each for $294.00
Description & Specifications

Specifications

Formula Weight 315.79
Chemical Name or Material 3,4,5-Tribromopyridine
Assay 97%
Molecular Formula C5H2Br3N
MDL Number MFCD01646053
CAS 2457-48-9
Melting Point 105° to 109°C
Health Hazard 1 Danger
Quantity 250mg
Health Hazard 2 GHS H Statement
H301-H311-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Danger

GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.