Learn More
3,4,5-Tribromo-1H-pyrazole, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1203114
Description
2,4,5-Tribromoimidazole(2,4,5-TBI) on condensation with sugar precursors yields 2,4,5-TBI nucleosides. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,4,5-Tribromoimidazole(2,4,5-TBI) on condensation with sugar precursors yields 2,4,5-TBI nucleosides. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Insoluble in water. Soluble in Chloroform.
Notes
Store in cool place. Keep away from oxidizing agents.
Specifications
3,4,5-Tribromo-1H-pyrazole | |
184°C to 188°C | |
C3HBr3N2 | |
25 g | |
3,4,5-tribromopyrazole, 1h-pyrazole, 3,4,5-tribromo, pyrazole, 3,4,5-tribromo, acmc-1c9yo, 3,4,5-tribromo pyrazole, ksc497i1h, #, 3,4,5-tribromo-1h-pyrazole 5g | |
TXQKCKQJBGFUBF-UHFFFAOYSA-N | |
3,4,5-tribromo-1H-pyrazole | |
627674 | |
97% |
17635-44-8 | |
Odorless | |
MFCD00040248 | |
113501 | |
Insoluble in water. Soluble in Chloroform. | |
C1(=C(NN=C1Br)Br)Br | |
304.767 | |
304.78 |
Chemical Identifiers
17635-44-8 | |
304.767 | |
TXQKCKQJBGFUBF-UHFFFAOYSA-N | |
627674 | |
C1(=C(NN=C1Br)Br)Br |
C3HBr3N2 | |
MFCD00040248 | |
3,4,5-tribromopyrazole, 1h-pyrazole, 3,4,5-tribromo, pyrazole, 3,4,5-tribromo, acmc-1c9yo, 3,4,5-tribromo pyrazole, ksc497i1h, #, 3,4,5-tribromo-1h-pyrazole 5g | |
3,4,5-tribromo-1H-pyrazole |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only