missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,4,5-Tribromo-1H-pyrazole, 97%

Manufacturer:  Alfa Aesar™ A1203122

 View more versions of this product

Catalog No. AAA1203122



Description

Description

Applications
2,4,5-Tribromoimidazole(2,4,5-TBI) on condensation with sugar precursors yields 2,4,5-TBI nucleosides. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water. Soluble in Chloroform.

Notes
Store in cool place. Keep away from oxidizing agents.
Specifications

Specifications

3,4,5-Tribromo-1H-pyrazole
184°C to 188°C
17635-44-8
MFCD00040248
3,4,5-tribromopyrazole, 1h-pyrazole, 3,4,5-tribromo, pyrazole, 3,4,5-tribromo, acmc-1c9yo, 3,4,5-tribromo pyrazole, ksc497i1h, #, 3,4,5-tribromo-1h-pyrazole 5g
C1(=C(NN=C1Br)Br)Br
304.767
304.78
Odorless
100g
C3HBr3N2
113501
TXQKCKQJBGFUBF-UHFFFAOYSA-N
3,4,5-tribromo-1H-pyrazole
627674
97%

Chemical Identifiers

17635-44-8
304.767
TXQKCKQJBGFUBF-UHFFFAOYSA-N
627674
C1(=C(NN=C1Br)Br)Br
C3HBr3N2
MFCD00040248
3,4,5-tribromopyrazole, 1h-pyrazole, 3,4,5-tribromo, pyrazole, 3,4,5-tribromo, acmc-1c9yo, 3,4,5-tribromo pyrazole, ksc497i1h, #, 3,4,5-tribromo-1h-pyrazole 5g
3,4,5-tribromo-1H-pyrazole
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents