missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3',4',5,7-Tetramethoxyflavone, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L14496MC

 View more versions of this product

Catalog No. AAL14496MC



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3',4',5,7-Tetramethoxyflavone
193°C to 197°C
MFCD00017558
336320
CLXVBVLQKLQNRQ-UHFFFAOYSA-N
2-(3,4-dimethoxyphenyl)-5,7-dimethoxychromen-4-one
631170
97%
855-97-0
C19H18O6
100 mg
3',4',5,7-tetramethoxyflavone, luteolin tetramethyl ether, tetramethoxyluteolin, 5,7,3',4'-tetramethylluteolin, tetramethylluteolin, 2-3,4-dimethoxyphenyl-5,7-dimethoxy-4h-chromen-4-one, luteolin tetramethylether, 2-3,4-dimethoxyphenyl-5,7-dimethoxychromen-4-one, luteolin 5,7,3',4'-tetramethyl ether, 4h-1-benzopyran-4-one, 2-3,4-dimethoxyphenyl-5,7-dimethoxy
COC1=CC(OC)=C2C(=O)C=C(OC2=C1)C1=CC=C(OC)C(OC)=C1
342.35
342.35

Chemical Identifiers

855-97-0
342.35
CLXVBVLQKLQNRQ-UHFFFAOYSA-N
631170
COC1=CC(OC)=C2C(=O)C=C(OC2=C1)C1=CC=C(OC)C(OC)=C1
C19H18O6
MFCD00017558
3',4',5,7-tetramethoxyflavone, luteolin tetramethyl ether, tetramethoxyluteolin, 5,7,3',4'-tetramethylluteolin, tetramethylluteolin, 2-3,4-dimethoxyphenyl-5,7-dimethoxy-4h-chromen-4-one, luteolin tetramethylether, 2-3,4-dimethoxyphenyl-5,7-dimethoxychromen-4-one, luteolin 5,7,3',4'-tetramethyl ether, 4h-1-benzopyran-4-one, 2-3,4-dimethoxyphenyl-5,7-dimethoxy
2-(3,4-dimethoxyphenyl)-5,7-dimethoxychromen-4-one
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only