missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)quinoline, TCI America™

Manufacturer:  TCI America T31805G

 View more versions of this product

Catalog No. T31805G



Specifications

Specifications

3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)quinoline
61°C
171364-85-5
MFCD05155218
ARRJAONCYUAPJR-UHFFFAOYSA-N
3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)quinoline
10848590
Crystal-Powder at 20°C
White-Yellow
5g
C15H18BNO2
3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl quinoline, quinoline-3-boronic acid pinacol ester, 3-quinolineboronic acid pinacol ester, quinoline-3-boronic acid pinacolate, 3-tetramethyl-1,3,2-dioxaborolan-2-yl quinoline, quinoline-3-boronic acid, pinacol ester, quinoline, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, pubchem1882, quinoline-3-boronic acidpinacolate, quinoline-3-boronicacidpinacolester
B1(OC(C(O1)(C)C)(C)C)C2=CC3=CC=CC=C3N=C2
255.124
255.12
≥98.0% (GC,T)

Chemical Identifiers

171364-85-5
255.124
ARRJAONCYUAPJR-UHFFFAOYSA-N
10848590
B1(OC(C(O1)(C)C)(C)C)C2=CC3=CC=CC=C3N=C2
C15H18BNO2
MFCD05155218
3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl quinoline, quinoline-3-boronic acid pinacol ester, 3-quinolineboronic acid pinacol ester, quinoline-3-boronic acid pinacolate, 3-tetramethyl-1,3,2-dioxaborolan-2-yl quinoline, quinoline-3-boronic acid, pinacol ester, quinoline, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, pubchem1882, quinoline-3-boronic acidpinacolate, quinoline-3-boronicacidpinacolester
3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)quinoline
Safety and Handling

Safety and Handling

TSCA : No

Documents