missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenol, 97%, ACROS Organics™

Chemical Identifiers

CAS 214360-76-6
Molecular Formula C12H17BO3
Molecular Weight (g/mol) 220.075
MDL Number MFCD02093755
InChI Key MUKIFYQKIZOYKT-UHFFFAOYSA-N
Synonym 2-3-hydroxyphenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
PubChem CID 2734623
IUPAC Name 3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC=C2)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC366410050 SDS View Documents Thermo Fisher Scientific
AC366410050
5g Glass Bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

214360-76-6
220.075
MUKIFYQKIZOYKT-UHFFFAOYSA-N
2734623
B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC=C2)O
C12H17BO3
MFCD02093755
2-3-hydroxyphenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol

Specifications

Authentic
Brown
5g
96.0
97%
MFCD02093755
Solubility in water: insoluble in water.
B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC=C2)O
220.075
220.08
97%
Glass Bottle
90.0°C to 96.0°C
214360-76-6
100.0
C12H17BO3
2-3-hydroxyphenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
MUKIFYQKIZOYKT-UHFFFAOYSA-N
3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol
2734623
Crystalline Powder
3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenol, 97%
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Recommended Storage : Refrigerator +4°C