Learn More
3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid, 97%, ACROS Organics™
Manufacturer: Fisher Scientific Acros AC367570050
Specifications
3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid | |
Glass bottle | |
206.0°C to 211.0°C | |
97% | |
96.0 | |
97% | |
MFCD03411930 | |
Solubility in water: insoluble in water. | |
B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC=C2)C(=O)O | |
248.09 | |
248.09 | |
97% |
Authentic | |
Beige to White | |
5g | |
269409-73-6 | |
100.0 | |
C13H17BO4 | |
3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoic acid, 3-carboxyphenyl boronic acid, pinacol ester, 3-carboxyphenylboronic acid pinacol ester, 3-carboxyphenylboronic acid, pinacol ester, 3-carboxyphenylboronicacidpinacolester, 3-cpbe, 3-tetramethyl-1,3,2-dioxaborolan-2-yl benzoic acid, amtb044, benzoic acid, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, pubchem17048 | |
OPWAPCOSDAFWFB-UHFFFAOYSA-N | |
3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid | |
2734653 | |
Crystalline Powder |
Chemical Identifiers
269409-73-6 | |
248.09 | |
OPWAPCOSDAFWFB-UHFFFAOYSA-N | |
2734653 | |
B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC=C2)C(=O)O |
C13H17BO4 | |
MFCD03411930 | |
3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoic acid, 3-carboxyphenyl boronic acid, pinacol ester, 3-carboxyphenylboronic acid pinacol ester, 3-carboxyphenylboronic acid, pinacol ester, 3-carboxyphenylboronicacidpinacolester, 3-cpbe, 3-tetramethyl-1,3,2-dioxaborolan-2-yl benzoic acid, amtb044, benzoic acid, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, pubchem17048 | |
3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
Recommended Storage : Refrigerator +4°C