Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-[3-(Trifluoromethyl)phenyl]propanol, 98%

Chemical Identifiers

Chemical Name or Material 3-[3-(Trifluoromethyl)phenyl]propanol
CAS 78573-45-2
Formula Weight 204.19
MDL Number MFCD08706408
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6455303

 
alfa aesar™
H6455303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6455306

 
alfa aesar™
H6455306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH64553MD

 
alfa aesar™
H64553MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 204.19
Boiling Point 68° to 71°C (0.4mmHg)
Physical Form Liquid
Density 1.10g/mL
Quantity 1g
Chemical Name or Material 3-[3-(Trifluoromethyl)phenyl]propanol
Assay 98%
MDL Number MFCD08706408
CAS 78573-45-2
Color Colorless