missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-[3-(Trifluoromethyl)phenyl]-1-propanol, TCI America™

Manufacturer:  TCI America T30955G

 View more versions of this product

Catalog No. T30955G



Specifications

Specifications

3-[3-(Trifluoromethyl)phenyl]-1-propanol
Yellow
78573-45-2
MFCD08706408
QWXKQVIMGVVIBX-UHFFFAOYSA-N
3-[3-(trifluoromethyl)phenyl]propan-1-ol
18755618
204.19
≥98.0% (GC)
60°C
5g
C10H11F3O
3-3-trifluoromethyl phenyl propan-1-ol, 3-3-trifluoromethyl phenyl propan-1-ol, 3-3'-trifluoromethyl phenyl propanol, 3-3-trifluoromethyl phenyl-1-propanol, 3-trifluoromethyl benzenepropanol, 3-3-trifluoromethyl-phenyl-propan-1-ol, 3-3'-trifluoromethylphenyl propanol, benzenepropanol, 3-trifluoromethyl, pubchem13744, benzenepropanol,3-trifluoromethyl
C1=CC(=CC(=C1)C(F)(F)F)CCCO
204.192
CHEBI:48528
Clear Liquid at 20°C

Chemical Identifiers

78573-45-2
204.192
QWXKQVIMGVVIBX-UHFFFAOYSA-N
18755618
3-[3-(trifluoromethyl)phenyl]propan-1-ol
C10H11F3O
MFCD08706408
3-3-trifluoromethyl phenyl propan-1-ol, 3-3-trifluoromethyl phenyl propan-1-ol, 3-3'-trifluoromethyl phenyl propanol, 3-3-trifluoromethyl phenyl-1-propanol, 3-trifluoromethyl benzenepropanol, 3-3-trifluoromethyl-phenyl-propan-1-ol, 3-3'-trifluoromethylphenyl propanol, benzenepropanol, 3-trifluoromethyl, pubchem13744, benzenepropanol,3-trifluoromethyl
CHEBI:48528
C1=CC(=CC(=C1)C(F)(F)F)CCCO
Safety and Handling

Safety and Handling

TSCA : No

Documents