Please login to your online account to display your discounted pricing

3-[3-(Trifluoromethyl)phenoxy]benzaldehyde, 97%, ACROS Organics™


Quantity 2.5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  187410025

 View more versions of this product

Catalog No. AC187410025

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Liquid
Formula Weight 266.21
Assay ≥96.0%
Quantity 2.5g
Color Clear yellow
Density 1.2800g/mL
CAS Max % 100.0
CAS Min % 96.0
Refractive Index 1.5365 to 1.5385 (20°C, 589nm)
Molecular Formula C14 H9 F3 O2
Infrared Spectrum Authentic
Packaging Glass bottle
Linear Formula CF3C6H4OC6H4CHO
Name Note 97%
Assay Percent Range 97%
MDL Number MFCD00003356
CAS 78725-46-9
Chemical Name or Material 3-[3-(Trifluoromethyl)phenoxy]benzaldehyde

Specifications

Chemical Name or Material 3-[3-(Trifluoromethyl)phenoxy]benzaldehyde
Name Note 97%
CAS 78725-46-9
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C14 H9 F3 O2
Linear Formula CF3C6H4OC6H4CHO
Formula Weight 266.21
MDL Number MFCD00003356