Please login to your online account to display your discounted pricing

3-[3-(Trifluoromethyl)phenoxy]benzaldehyde, 97%, ACROS Organics™

$87.70 - $87.70

Chemical Identifiers

Chemical Name or Material 3-[3-(Trifluoromethyl)phenoxy]benzaldehyde
Name Note 97%
CAS 78725-46-9
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C14 H9 F3 O2
Linear Formula CF3C6H4OC6H4CHO
Formula Weight 266.21
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC187410025

  SDS
thermo fisher scientific
187410025
Glass bottle 2.5g Each for $87.70
Description & Specifications

Specifications

Physical Form Liquid
Formula Weight 266.21
Assay ≥96.0%
Quantity 2.5g
Color Clear yellow
Density 1.2800g/mL
CAS Max % 100.0
CAS Min % 96.0
Refractive Index 1.5365 to 1.5385 (20°C, 589nm)
Molecular Formula C14 H9 F3 O2
Infrared Spectrum Authentic
Packaging Glass bottle
Linear Formula CF3C6H4OC6H4CHO
Name Note 97%
Assay Percent Range 97%
MDL Number MFCD00003356
CAS 78725-46-9
Chemical Name or Material 3-[3-(Trifluoromethyl)phenoxy]benzaldehyde