Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-[3-(Trifluoromethoxy)phenyl]propionic acid, 95%

$115.12 - $325.46

Chemical Identifiers

Chemical Name or Material 3-[3-(Trifluoromethoxy)phenyl]propionic acid
CAS 168833-77-0
Molecular Formula C10H9F3O3
Formula Weight 234.17
MDL Number MFCD01317834
Synonym 3-(Trifluoromethoxy)hydrocinnamic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2123301

  SDS
alfa aesar™
B2123301
0.25g Each for $115.12

AAB2123303

  SDS
alfa aesar™
B2123303
1g Each for $325.46
Description & Specifications

Specifications

Formula Weight 234.17
Chemical Name or Material 3-[3-(Trifluoromethoxy)phenyl]propionic acid
Assay 95%
Molecular Formula C10H9F3O3
MDL Number MFCD01317834
CAS 168833-77-0
Synonym 3-(Trifluoromethoxy)hydrocinnamic acid
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 0.25g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.