missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,3'-Thiodipropionic acid, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 138750250

 View more versions of this product

Catalog No. AC138750250



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3,3′-Thiodipropionic acid
111-17-1
100.0
White
Authentic
Plastic bottle
S(CH2CH2CO2H)2
25 g
15, 9485
Solubility in water: 40g/L (25°C). Other solubilities: freely soluble in hot water, freely soluble in alcohol and acetone
C(CSCCC(=O)O)C(=O)O
178.2
178.2
Fine Crystalline Powder
99%
98.5
130.0°C to 133.0°C
128°C
99%
C6H10O4S
MFCD00002781
03, IV, 735
3,3'-thiodipropionic acid, thiodipropionic acid, propanoic acid, 3,3'-thiobis, bis 2-carboxyethyl sulfide, tyox a, thiodihydracrylic acid, 4-thiaheptanedioic acid, tdpa, sulfide, bis 2-carboxyethyl, 3,3'-thiodipropanoic acid
ODJQKYXPKWQWNK-UHFFFAOYSA-N
3-(2-carboxyethylsulfanyl)propanoic acid
8096
99%

Chemical Identifiers

111-17-1
178.2
ODJQKYXPKWQWNK-UHFFFAOYSA-N
8096
C(CSCCC(=O)O)C(=O)O
C6H10O4S
MFCD00002781
3,3'-thiodipropionic acid, thiodipropionic acid, propanoic acid, 3,3'-thiobis, bis 2-carboxyethyl sulfide, tyox a, thiodihydracrylic acid, 4-thiaheptanedioic acid, tdpa, sulfide, bis 2-carboxyethyl, 3,3'-thiodipropanoic acid
3-(2-carboxyethylsulfanyl)propanoic acid
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER

GHS Signal Word: Warning

EINECSNumber : 203-841-3

RTECSNumber : UF7990000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only