Please login to your online account to display your discounted pricing

3,3'-Thiodipropionic Acid 99%, ACROS Organics™


Quantity 5g
Packaging Glass Bottle

Manufacturer: thermo fisher scientific  138750050

 View more versions of this product

Catalog No. AC138750050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Fine Crystalline Powder
Formula Weight 178.2
Quantity 5g
Color White
Beilstein 03, IV, 735
Merck Index 15, 9485
Packaging Glass Bottle
EINECS Number 203-841-3
TSCA TSCA
Assay 98.5 to 101.5%
RTECS Number UF7990000
Melting Point 130° to 133°C
CAS Max % 100.0
Chemical Name or Material 3, 3';-Thiodipropionic acid
Infrared Spectrum Authentic
Name Note 99%
Assay Percent Range 99%
Molecular Formula C6H10O4S
Linear Formula S(CH2CH2CO2H)2
CAS Min % 98.5
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00002781
CAS 111-17-1

Specifications

Chemical Name or Material 3, 3';-Thiodipropionic acid
Name Note 99%
CAS 111-17-1
CAS Min % 98.5
CAS Max % 100.0
Assay 98.5 to 101.5%
Assay Percent Range 99%
Molecular Formula C6H10O4S
Linear Formula S(CH2CH2CO2H)2
Formula Weight 178.2
MDL Number MFCD00002781
Accessories

Warning

GHS P Statement:
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

SDS & Safety Documents