Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,3-Tetramethyleneglutarimide, 98%

Chemical Identifiers

Chemical Name or Material 3,3-Tetramethyleneglutarimide
CAS 1075-89-4
Molecular Formula C9H13NO2
Formula Weight 167.21
MDL Number MFCD00023871
UN Number UN2811
Beilstein 383702
Synonym 8-Azaspiro[4.5]decane-7,9-dione; 1,1-Cyclopentanediacetimide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2486406

  SDS
alfa aesar™
B2486406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2486414

  SDS
alfa aesar™
B2486414
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 167.21
Odor Odorless
UN Number UN2811
Beilstein 383702
Health Hazard 3 GHS P Statement
P260-P280f-P273-P309-P310
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective clothing.
Avoid release to the environment.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Assay 98%
Melting Point 153° to 156°C
Molecular Formula C9H13NO2
Health Hazard 2 GHS H Statement
H301
Toxic if swallowed.
MDL Number MFCD00023871
CAS 1075-89-4
Chemical Name or Material 3,3-Tetramethyleneglutarimide
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Health Hazard 1 Danger
Quantity 5g
Synonym 8-Azaspiro[4.5]decane-7,9-dione; 1,1-Cyclopentanediacetimide
Safety and Handling

Danger

GHS P Statement
P260-P280f-P273-P309-P310
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective clothing.
Avoid release to the environment.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H301
Toxic if swallowed.