Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,3-Tetramethyleneglutaric acid, 98+%

$98.39 - $363.14

Chemical Identifiers

Chemical Name or Material 3,3-Tetramethyleneglutaric acid
CAS 16713-66-9
Molecular Formula C9H14O4
Formula Weight 186.21
MDL Number MFCD00001388
Beilstein 2099523
Synonym 1,1-Cyclopentanediacetic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2014614

  SDS
alfa aesar™
B2014614
25g Each for $98.39

AAB2014622

  SDS
alfa aesar™
B2014622
100g Each for $363.14
Description & Specifications

Specifications

Formula Weight 186.21
Health Hazard 1 Warning
Synonym 1,1-Cyclopentanediacetic acid
Molecular Formula C9H14O4
Quantity 25g
Beilstein 2099523
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 179° to 181°C
MDL Number MFCD00001388
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 3,3-Tetramethyleneglutaric acid
Assay ≥98%
CAS 16713-66-9
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.