missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(3-Pyridyl)-L-alanine, 98%

Manufacturer:  Alfa Aesar™ H6394106

 View more versions of this product

Catalog No. AAH6394106



Description

Description

Applications
3-(3-Pyridyl)-L-alanine is a antimycobacterial cyclodepsipeptide. PyrA is used to activate other aromatic acids.

Solubility
Soluble in clear solution (0.3 gram in 2 ml DMF).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Specifications

3-(3-Pyridyl)-L-alanine
C8H10N2O2
4351583
DFZVZEMNPGABKO-ZETCQYMHSA-N
(2S)-2-amino-3-pyridin-3-ylpropanoic acid
152953
Powder
White
5g
64090-98-8
MFCD00079676
3-3-pyridyl-l-alanine, l-3-pyridylalanine, s-2-amino-3-pyridin-3-yl propanoic acid, h-3-pal-oh, 3-3-pyridyl alanine, 2s-2-amino-3-pyridin-3-yl propanoic acid, l-3-3-pyridyl-alanine, 3'-pyridyl-l-ala, h-beta-3-pyridyl-ala-oh, 2s-2-amino-3-pyridin-3-ylpropanoic acid
C1=CC(=CN=C1)CC(C(=O)O)N
166.18
166.18
98%
248°C to 252°C (decomposition)

Chemical Identifiers

64090-98-8
166.18
DFZVZEMNPGABKO-ZETCQYMHSA-N
152953
C1=CC(=CN=C1)CC(C(=O)O)N
C8H10N2O2
MFCD00079676
3-3-pyridyl-l-alanine, l-3-pyridylalanine, s-2-amino-3-pyridin-3-yl propanoic acid, h-3-pal-oh, 3-3-pyridyl alanine, 2s-2-amino-3-pyridin-3-yl propanoic acid, l-3-3-pyridyl-alanine, 3'-pyridyl-l-ala, h-beta-3-pyridyl-ala-oh, 2s-2-amino-3-pyridin-3-ylpropanoic acid
(2S)-2-amino-3-pyridin-3-ylpropanoic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents