Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(3-Pyridyl)benzeneboronic acid, 98%

Chemical Identifiers

Chemical Name or Material 3-(3-Pyridyl)benzeneboronic acid
CAS 351422-72-5
Formula Weight 199.02
MDL Number MFCD16658918
Beilstein 20520996
Synonym 3-(3-Pyridyl)phenylboronic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6473903

  SDS
alfa aesar™
H6473903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6473906

  SDS
alfa aesar™
H6473906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH64739MD

  SDS
alfa aesar™
H64739MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 199.02
Chemical Name or Material 3-(3-Pyridyl)benzeneboronic acid
Physical Form Powder
Assay 98%
MDL Number MFCD16658918
CAS 351422-72-5
Beilstein 20520996
Synonym 3-(3-Pyridyl)phenylboronic acid
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Color Off-white
Quantity 1g

Applications
It is used as an OLED material. Phenylboronic acids are efficient phenylating agents in copper promoted CN and CO bond cross-coupling reactions.

Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.