Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,3-Pentamethylene glutarimide, ≥98%

Chemical Identifiers

Chemical Name or Material 3,3-Pentamethylene glutarimide
CAS 1130-32-1
Formula Weight 181.23
MDL Number MFCD00023872
Beilstein 383782
Synonym 3-Azaspiro[5.5]undecane-2,4-dione
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6490203

 
alfa aesar™
H6490203
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6490206

 
alfa aesar™
H6490206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6490214

 
alfa aesar™
H6490214
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 168° to 170°C
Formula Weight 181.23
Chemical Name or Material 3,3-Pentamethylene glutarimide
Physical Form Solid
Assay ≥98%
Quantity 1g
MDL Number MFCD00023872
CAS 1130-32-1
Beilstein 383782
Synonym 3-Azaspiro[5.5]undecane-2,4-dione
Storage Note 1 -30° to -10°C
RTECS Number CM0215000
Safety and Handling

-30° to -10°C