missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(3-Oxycyclohexyl)indole, 95%

$172.38 - $410.41

Chemical Identifiers

CAS 935758-78-4
Synonym 3-(3-Indolyl)cyclohexanone
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH63476MD
View Documents
Alfa Aesar™
H63476MD
250mg Each for $172.38
 
AAH6347603
View Documents
Alfa Aesar™
H6347603
1g Each for $410.41
 
Specifications

Chemical Identifiers

935758-78-4
3-(3-Indolyl)cyclohexanone

Specifications

250mg
7639563
213.28
95%
935758-78-4
3-(3-Indolyl)cyclohexanone
Solid
3-(3-Oxycyclohexyl)indole
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures