missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,3′-Methylenediindole, TCI America™

$70.28 - $206.25

Chemical Identifiers

CAS 1968-05-4
Molecular Formula C17H14N2
Molecular Weight (g/mol) 246.31
MDL Number MFCD00195766
InChI Key VFTRKSBEFQDZKX-UHFFFAOYSA-N
Synonym 3,3'-diindolylmethane, di 1h-indol-3-yl methane, diindolylmethane, 3,3'-methylenediindole, 3,3'-methylenebis-1h-indole, 1h-indole, 3,3'-methylenebis, dim, 3-diindolyl methane, arundine, 3,3'-methylenebis 1h-indole
PubChem CID 3071
ChEBI CHEBI:50182
IUPAC Name 3-(1H-indol-3-ylmethyl)-1H-indole
SMILES C1=CC=C2C(=C1)C(=CN2)CC3=CNC4=CC=CC=C43
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M13995G View Documents TCI America
M13995G
5g Each for $70.28
 
M139925G View Documents TCI America
M139925G
25g Each for $206.25
 
Specifications

Chemical Identifiers

1968-05-4
246.31
VFTRKSBEFQDZKX-UHFFFAOYSA-N
3071
3-(1H-indol-3-ylmethyl)-1H-indole
C17H14N2
MFCD00195766
3,3'-diindolylmethane, di 1h-indol-3-yl methane, diindolylmethane, 3,3'-methylenediindole, 3,3'-methylenebis-1h-indole, 1h-indole, 3,3'-methylenebis, dim, 3-diindolyl methane, arundine, 3,3'-methylenebis 1h-indole
CHEBI:50182
C1=CC=C2C(=C1)C(=CN2)CC3=CNC4=CC=CC=C43

Specifications

Undesignated
5g
C17H14N2
3,3'-diindolylmethane, di 1h-indol-3-yl methane, diindolylmethane, 3,3'-methylenediindole, 3,3'-methylenebis-1h-indole, 1h-indole, 3,3'-methylenebis, dim, 3-diindolyl methane, arundine, 3,3'-methylenebis 1h-indole
C1=CC=C2C(=C1)C(=CN2)CC3=CNC4=CC=CC=C43
246.31
CHEBI:50182
Crystal-Powder at 20°C
3,3′-Methylenediindole
167°C
1968-05-4
MFCD00195766
VFTRKSBEFQDZKX-UHFFFAOYSA-N
3-(1H-indol-3-ylmethyl)-1H-indole
3071
246.31
≥98.0% (N)
Safety and Handling

Safety and Handling

TSCA : No