missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,3′-Methylenebis[1-(2,6-diisopropylphenyl)-3-imidazolium Bromide], TCI America™

$345.28 - $1,034.33

Chemical Identifiers

CAS 889617-36-1
Molecular Formula C31H42Br2N4
Molecular Weight (g/mol) 630.513
InChI Key ZZGWEIDPDBDPOH-UHFFFAOYSA-L
PubChem CID 78198750
IUPAC Name 1-[2,6-di(propan-2-yl)phenyl]-3-[[3-[2,6-di(propan-2-yl)phenyl]imidazol-1-ium-1-yl]methyl]imidazol-3-ium;dibromide
SMILES CC(C)C1=C(C(=CC=C1)C(C)C)N2C=C[N+](=C2)C[N+]3=CN(C=C3)C4=C(C=CC=C4C(C)C)C(C)C.[Br-].[Br-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M27915G View Documents TCI America
M27915G
5g Each for $1,034.33
 
M27911G View Documents TCI America
M27911G
1g Each for $345.28
 
Specifications

Chemical Identifiers

889617-36-1
630.513
78198750
CC(C)C1=C(C(=CC=C1)C(C)C)N2C=C[N+](=C2)C[N+]3=CN(C=C3)C4=C(C=CC=C4C(C)C)C(C)C.[Br-].[Br-]
C31H42Br2N4
ZZGWEIDPDBDPOH-UHFFFAOYSA-L
1-[2,6-di(propan-2-yl)phenyl]-3-[[3-[2,6-di(propan-2-yl)phenyl]imidazol-1-ium-1-yl]methyl]imidazol-3-ium;dibromide

Specifications

White
5g
C31H42Br2N4
CC(C)C1=C(C(=CC=C1)C(C)C)N2C=C[N+](=C2)C[N+]3=CN(C=C3)C4=C(C=CC=C4C(C)C)C(C)C.[Br-].[Br-]
630.513
630.51
≥98.0% (HPLC,T)
277°C
889617-36-1
ZZGWEIDPDBDPOH-UHFFFAOYSA-L
1-[2,6-di(propan-2-yl)phenyl]-3-[[3-[2,6-di(propan-2-yl)phenyl]imidazol-1-ium-1-yl]methyl]imidazol-3-ium;dibromide
78198750
Crystal-Powder at 20°C
3,3′-Methylenebis[1-(2,6-diisopropylphenyl)-3-imidazolium Bromide]
Safety and Handling

Safety and Handling

TSCA : No