missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(3-Methyl-2-pyridyl)-5-phenyl-1,2,4-oxadiazole, 97%

Manufacturer:  Alfa Aesar™ H5467606

 View more versions of this product

Catalog No. AAH5467606

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

3-(3-Methyl-2-pyridyl)-5-phenyl-1,2,4-oxadiazole
1400191-51-6
MFCD20265320
ZHKWMFUDVLEAHG-UHFFFAOYSA-N
3-(3-methylpyridin-2-yl)-5-phenyl-1,2,4-oxadiazole
73995920
97%
5g
C14H11N3O
3-3-methyl-2-pyridyl-5-phenyl-1,2,4-oxadiazole
CC1=C(N=CC=C1)C2=NOC(=N2)C3=CC=CC=C3
237.262
237.26

Chemical Identifiers

1400191-51-6
237.262
ZHKWMFUDVLEAHG-UHFFFAOYSA-N
73995920
CC1=C(N=CC=C1)C2=NOC(=N2)C3=CC=CC=C3
C14H11N3O
MFCD20265320
3-3-methyl-2-pyridyl-5-phenyl-1,2,4-oxadiazole
3-(3-methylpyridin-2-yl)-5-phenyl-1,2,4-oxadiazole
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents