missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(3-Methyl-2-pyridyl)-5-phenyl-1,2,4-oxadiazole, 97%

Manufacturer:  Alfa Aesar™ H5467603

 View more versions of this product

Catalog No. AAH5467603

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

3-(3-Methyl-2-pyridyl)-5-phenyl-1,2,4-oxadiazole
1400191-51-6
MFCD20265320
ZHKWMFUDVLEAHG-UHFFFAOYSA-N
3-(3-methylpyridin-2-yl)-5-phenyl-1,2,4-oxadiazole
73995920
97%
1g
C14H11N3O
3-3-methyl-2-pyridyl-5-phenyl-1,2,4-oxadiazole
CC1=C(N=CC=C1)C2=NOC(=N2)C3=CC=CC=C3
237.262
237.26

Chemical Identifiers

1400191-51-6
237.262
ZHKWMFUDVLEAHG-UHFFFAOYSA-N
73995920
CC1=C(N=CC=C1)C2=NOC(=N2)C3=CC=CC=C3
C14H11N3O
MFCD20265320
3-3-methyl-2-pyridyl-5-phenyl-1,2,4-oxadiazole
3-(3-methylpyridin-2-yl)-5-phenyl-1,2,4-oxadiazole
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents