missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(3-Methyl-1,2,4-oxadiazol-5-yl)piperidine, 98%

$104.53 - $363.27

Chemical Identifiers

CAS 21767-12-4
Molecular Formula C8H13N3O
Formula Weight 167.21
MDL Number MFCD07801211
Chemical Name or Material 3-(3-Methyl-1,2,4-oxadiazol-5-yl)piperidine
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5088303 SDS Alfa Aesar™
H5088303
1g Each for $363.27
AAH50883MD SDS Alfa Aesar™
H50883MD
250mg Each for $104.53
Specifications Description & Specifications

Chemical Identifiers

21767-12-4
167.21
3-(3-Methyl-1,2,4-oxadiazol-5-yl)piperidine
C8H13N3O
MFCD07801211

Specifications

21767-12-4
167.21
1g
98%
C8H13N3O
MFCD07801211
3-(3-Methyl-1,2,4-oxadiazol-5-yl)piperidine

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning