Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(3-Methoxyphenoxy)piperidine, 97%

$91.46 - $331.49

Chemical Identifiers

Chemical Name or Material 3-(3-Methoxyphenoxy)piperidine
CAS 902837-25-6
Molecular Formula C12H17NO2
Formula Weight 207.27
MDL Number MFCD08060989
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5079403

  SDS
alfa aesar™
H5079403
1g Each for $331.49

AAH50794MD

  SDS
alfa aesar™
H50794MD
250mg Each for $91.46
Description & Specifications

Specifications

Formula Weight 207.27
Health Hazard 1 Warning
Chemical Name or Material 3-(3-Methoxyphenoxy)piperidine
CAS 902837-25-6
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 97%
MDL Number MFCD08060989
Quantity 1g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C12H17NO2

Applications
It is employed as intermediate for pharmaceutical.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.