Please login to your online account to display your discounted pricing

3-(3-Isocyanatophenyl)-5-methyl-1,2,4-oxadiazole, 97%, Maybridge


Quantity 250mg
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC39106CB

 View more versions of this product

Catalog No. CC39106CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 201.18
Quantity 250mg
Assay 97%
Molecular Formula C10H7N3O2
CAS 852180-69-9
Chemical Name or Material 3-(3-Isocyanatophenyl)-5-methyl-1, 2, 4-oxadiazole
Packaging Amber Glass Bottle
Assay Percent Range 97%

Specifications

Chemical Name or Material 3-(3-Isocyanatophenyl)-5-methyl-1, 2, 4-oxadiazole
Assay 97%
Assay Percent Range 97%
Molecular Formula C10H7N3O2
Formula Weight 201.18