Please login to your online account to display your discounted pricing

3-(3-Isocyanatophenyl)-5-methyl-1,2,4-oxadiazole, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 3-(3-Isocyanatophenyl)-5-methyl-1, 2, 4-oxadiazole
Assay 97%
Assay Percent Range 97%
Molecular Formula C10H7N3O2
Formula Weight 201.18
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

CC39106CB

  SDS
fisher scientific company
CC39106CB
Amber Glass Bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CC39106DA

  SDS
fisher scientific company
CC39106DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 201.18
Quantity 250mg
Assay 97%
Chemical Name or Material 3-(3-Isocyanatophenyl)-5-methyl-1, 2, 4-oxadiazole
Molecular Formula C10H7N3O2
CAS 852180-69-9
Assay Percent Range 97%
Packaging Amber Glass Bottle